ContactUsLinkComponent
Quartz-Glass therm., +400+750:2°C, 450x8-9mm, cap. colourless, gallium filled, strip bearing the scale: quartz, total immersion, pin -top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARQ134016-GA
Supplier:  Amarell
Quartz-Glass therm., +400+750:2°C, 450x8-9mm, cap. colourless, gallium filled, strip bearing the scale: quartz, total immersion, pin -top finish with button, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARQ134016-GA
Supplier:  Amarell