ContactUsLinkComponent
API Gravity hydrometer ASTM, 1H-62, -1+11:0,1API, ref. temp. 60F, without therm., 330mm long 1 * 1 items
Catalog # AMARH825000
Supplier:  Amarell
API Gravity hydrometer ASTM, 1H-62, -1+11:0,1API, ref. temp. 60F, without therm., 330mm long 1 * 1 items
Catalog # AMARH825000
Supplier:  Amarell