ContactUsLinkComponent
API Gravity hydrometer ASTM, 35H-62, 70+76:0,1API, ref. temp. 60F, without therm., 163mm long 1 * 1 items
Catalog # AMARH825148
Supplier:  Amarell
API Gravity hydrometer ASTM, 35H-62, 70+76:0,1API, ref. temp. 60F, without therm., 163mm long 1 * 1 items
Catalog # AMARH825148
Supplier:  Amarell