ContactUsLinkComponent
API Gravity hydrometer ASTM, 25H-62, 20+26:0,1API, ref. temp. 60F, without therm., 163mm long 1 * 1 items
  AMARH825128
 :  Amarell
API Gravity hydrometer ASTM, 25H-62, 20+26:0,1API, ref. temp. 60F, without therm., 163mm long 1 * 1 items
  AMARH825128
 :  Amarell