ContactUsLinkComponent
Thermometer similar to IP 77 C, solid stem, white backed, -2+52:0,2C, red special liquid, immersion 100mm, 395x6-8mm, durable pigme nt, suitable for government calibration 1 * 1 items
Catalog # AMARA341080-FL
Supplier:  Amarell
Thermometer similar to IP 77 C, solid stem, white backed, -2+52:0,2C, red special liquid, immersion 100mm, 395x6-8mm, durable pigme nt, suitable for government calibration 1 * 1 items
Catalog # AMARA341080-FL
Supplier:  Amarell