ContactUsLinkComponent
Thermometer, solid stem, similar to IP 74 C, white backed, -35+70:0,5C, red special liquid, 61mm immersion, 310mm long, null
Catalog # AMARA341050-FL
Supplier:  Amarell
Thermometer, solid stem, similar to IP 74 C, white backed, -35+70:0,5C, red special liquid, 61mm immersion, 310mm long, null
Catalog # AMARA341050-FL
Supplier:  Amarell