ContactUsLinkComponent
Set for dropping point, DIN 51801 Bl. 1, consisting of: 2 therm. similar ASTM 2 C gallium filled, metal ferrule, test tube, metal ro d, corks, suitable for government verification 1 * 1 items
Catalog # AMARM234560-SET
Supplier:  Amarell
Set for dropping point, DIN 51801 Bl. 1, consisting of: 2 therm. similar ASTM 2 C gallium filled, metal ferrule, test tube, metal ro d, corks, suitable for government verification 1 * 1 items
Catalog # AMARM234560-SET
Supplier:  Amarell