ContactUsLinkComponent
Thermometer ähnlich IP 53 C, solid stem, white backed, 0+80:0,5°C, capillary tube specially coated inside, with non-wetting blue sp ecial liquid, durable pigment, null
Catalog # AMARA340780-CB
Supplier:  Amarell
Thermometer ähnlich IP 53 C, solid stem, white backed, 0+80:0,5°C, capillary tube specially coated inside, with non-wetting blue sp ecial liquid, durable pigment, null
Catalog # AMARA340780-CB
Supplier:  Amarell