ContactUsLinkComponent
Thermometer similar to IP 43 C, solid stem, white backed, 10+110:0,5°C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA340600-FL
Supplier:  Amarell
Thermometer similar to IP 43 C, solid stem, white backed, 10+110:0,5°C, red special liquid, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA340600-FL
Supplier:  Amarell