ContactUsLinkComponent
Hydrometer DIN 12791/BS 718, L50, 1,95-2,00:0,0005g/cm , ref. temp. 20C, without therm., max. 335mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH806254
Supplier:  Amarell
Hydrometer DIN 12791/BS 718, L50, 1,95-2,00:0,0005g/cm , ref. temp. 20C, without therm., max. 335mm long, suitable for government v erification 1 * 1 items
Catalog # AMARH806254
Supplier:  Amarell