ContactUsLinkComponent
Density hydrometer, 1,000-1,500:0,005g/cm , ref. temp. 20C, with therm., 0+40:1C, 280-350mm long, special liquid 1 * 1 items
Catalog # AMARH801682-FL
Supplier:  Amarell
Density hydrometer, 1,000-1,500:0,005g/cm , ref. temp. 20C, with therm., 0+40:1C, 280-350mm long, special liquid 1 * 1 items
Catalog # AMARH801682-FL
Supplier:  Amarell