ContactUsLinkComponent
Refill therm. for well scoop therm., -5+35:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue s pecial liquid, total immersion, null
Catalog # AMARK196250-CB
Supplier:  Amarell
Refill therm. for well scoop therm., -5+35:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue s pecial liquid, total immersion, null
Catalog # AMARK196250-CB
Supplier:  Amarell