0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Density hydrometer, 1,720-1,780:0,001g/cm , ref. temp. 20C, with therm., 350mm long, special liquid 1 * 1 items
AMARH800278-FL
:
Amarell
Density hydrometer, 1,720-1,780:0,001g/cm , ref. temp. 20C, with therm., 350mm long, special liquid 1 * 1 items
AMARH800278-FL
:
Amarell