ContactUsLinkComponent
Density hydrometer, 1,600-1,660:0,001g/cm , ref. temp. 20C, with therm., 350mm long, special liquid 1 * 1 items
Catalog # AMARH800274-FL
Supplier:  Amarell
Density hydrometer, 1,600-1,660:0,001g/cm , ref. temp. 20C, with therm., 350mm long, special liquid 1 * 1 items
Catalog # AMARH800274-FL
Supplier:  Amarell