Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Wine alcoholometer, 0-25Vol.%, without therm., ref. temp. 20C, 180mm long 1 * 1 items
Catalog # AMARH850320
Supplier:
Amarell
Expand image
Wine alcoholometer, 0-25Vol.%, without therm., ref. temp. 20C, 180mm long 1 * 1 items
Catalog # AMARH850320
Supplier:
Amarell