ContactUsLinkComponent
Distillation therm., similar to DIN 12779, 0+100:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, total immersio n, length 210mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259602-FL
Supplier:  Amarell
Distillation therm., similar to DIN 12779, 0+100:1°C, enclosed scale, cap. prismatic, colourless, red special liquid, total immersio n, length 210mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259602-FL
Supplier:  Amarell