ContactUsLinkComponent
Therm. for distillation range, similar to DIN 12785, solid stem, similar to ASTM 8 C (BST 1/-2/400), -2+400:1°C, length 385mm, white backed, gallium filled, total immesion, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARD258402-GA
Supplier:  Amarell
Therm. for distillation range, similar to DIN 12785, solid stem, similar to ASTM 8 C (BST 1/-2/400), -2+400:1°C, length 385mm, white backed, gallium filled, total immesion, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARD258402-GA
Supplier:  Amarell