ContactUsLinkComponent
Thermometer similar to AFNOR, STL/0,2/-15/45, solid stem, white backed, -15+45:0,2C, capillary tube specially coated inside, with non -wetting blue special liquid, 375mm long, total immersion, durable pigment, null
Catalog # AMARA411036-CB
Supplier:  Amarell
Thermometer similar to AFNOR, STL/0,2/-15/45, solid stem, white backed, -15+45:0,2C, capillary tube specially coated inside, with non -wetting blue special liquid, 375mm long, total immersion, durable pigment, null
Catalog # AMARA411036-CB
Supplier:  Amarell