ContactUsLinkComponent
Alcoholometer, similar to EUROPA class II, 60-70:0,1%vol., with therm. +5+25:0,2C with red or blue special liquid, 400mm long, suit able for government calibration 1 * 1 items
Catalog # AMARH848172-FL
Supplier:  Amarell
Alcoholometer, similar to EUROPA class II, 60-70:0,1%vol., with therm. +5+25:0,2C with red or blue special liquid, 400mm long, suit able for government calibration 1 * 1 items
Catalog # AMARH848172-FL
Supplier:  Amarell