ContactUsLinkComponent
Alcoholometer, EUROPA, class I, 0-10:0,1%vol., without therm., ref. temp. 20C, 400mm long, suitable for government verification 1 * 1 items
Catalog # AMARH848020
Supplier:  Amarell
Alcoholometer, EUROPA, class I, 0-10:0,1%vol., without therm., ref. temp. 20C, 400mm long, suitable for government verification 1 * 1 items
Catalog # AMARH848020
Supplier:  Amarell