ContactUsLinkComponent
Beer hydrometer, 1,0134-1,0276:0,0002g/cm , with therm. +13+26:0,2°C with red or blue special liquid, with correction scale, reading above, ref. temp. 20°C, 415mm long 1 * 1 items
Catalog # AMARH847504-FL
Supplier:  Amarell
Beer hydrometer, 1,0134-1,0276:0,0002g/cm , with therm. +13+26:0,2°C with red or blue special liquid, with correction scale, reading above, ref. temp. 20°C, 415mm long 1 * 1 items
Catalog # AMARH847504-FL
Supplier:  Amarell