ContactUsLinkComponent
Thermometer similar to AFNOR, STL/0,1/-25/5, -25+5:0,1°C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 375mm long, total immersion, durable pigment, null
Catalog # AMARA411000-CB
Supplier:  Amarell
Thermometer similar to AFNOR, STL/0,1/-25/5, -25+5:0,1°C, solid stem, white backed, capillary tube specially coated inside, with no n-wetting blue special liquid, 375mm long, total immersion, durable pigment, null
Catalog # AMARA411000-CB
Supplier:  Amarell