ContactUsLinkComponent
Thermometer similar to BS 593, F275C/100, 223+277:0,2°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, sui table for government calibration 1 * 1 items
  AMARA403398-GA
 :  Amarell
Thermometer similar to BS 593, F275C/100, 223+277:0,2°C, solid stem, white backed, gallium filled, 395mm long, 100mm immersion, sui table for government calibration 1 * 1 items
  AMARA403398-GA
 :  Amarell