ContactUsLinkComponent
Pocket saccharimeter, 14-21:0,1%mas., with therm. +10+25:0,2°C with red or blue special liquid, with correction scale, reading above , ref. temp. 20°C, 275mm long, suitable for government calibration 1 * 1 items
  AMARH847484-FL
 :  Amarell
Pocket saccharimeter, 14-21:0,1%mas., with therm. +10+25:0,2°C with red or blue special liquid, with correction scale, reading above , ref. temp. 20°C, 275mm long, suitable for government calibration 1 * 1 items
  AMARH847484-FL
 :  Amarell