ContactUsLinkComponent
Thermometer similar to BS 593, A40C/100, 0+40:0,1C, solid stem, white backed, red special liquid, 400mm long, 100mm immersion, dura ble pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA403244-FL
Supplier:  Amarell
Thermometer similar to BS 593, A40C/100, 0+40:0,1C, solid stem, white backed, red special liquid, 400mm long, 100mm immersion, dura ble pigment, suitable for government calibration 1 * 1 items
Catalog # AMARA403244-FL
Supplier:  Amarell