ContactUsLinkComponent
Saccharometer, simple type, 0-50:1/1B, without therm., ref. temp. 15C, 150mm long, in cardboard-case 1 * 1 items
Catalog # AMARH847050
Supplier:  Amarell
Saccharometer, simple type, 0-50:1/1B, without therm., ref. temp. 15C, 150mm long, in cardboard-case 1 * 1 items
Catalog # AMARH847050
Supplier:  Amarell