Thermometer similar to BS 1365, SB175C/total, 165+175:0,1°C, solid stem, white backed, gallium filled, 235mm long, total immersion, durable pigment, suitable for government calibration 1 * 1 items
Thermometer similar to BS 1365, SB175C/total, 165+175:0,1°C, solid stem, white backed, gallium filled, 235mm long, total immersion, durable pigment, suitable for government calibration 1 * 1 items