Order Entry
Export
Orders LinkContactUsLinkComponent
 

Scraper, Cuinox

Scraper, white, with red handle, 410 mm
  KINGEL605177
Scraper, Cuinox
  KINGEL605177
Scraper, Cuinox
Scraper, Cuinox

Scraper, white, with red handle, 410 mm

 
37,50 €
   
1 * 1 items
 

Heat resistant scraper with silicone blade and PE handle.

  • Heat resistant up to 260 °C
  • Ideal for use in kitchens and bakeries
 
  • Description:
    Scraper, white, with red handle
  • Length:
    410 mm
  • Seq:
    02