Inspection date labels
Inspection date labels, Vinyl, 2024, Brown, 15 mm
Catalog # BRDY834106
Supplier: Brady
Jump to:
Product Details & Documents
These inspection date labels are made of vinyl cloth. They can be used wherever a frequent and regular inspection of machinery and equipment is required. Simply mark the label with a hole punch and apply wherever you need it.
- Show evidence of unauthorised removal
Use the yearly colour system to easily identify when your equipment was last inspected.
Documents
Specifications
- Colour:Brown
- Description:Inspection date labels
- Dimensions:15 mm
- Material:Vinyl
- Legend:2024