Various replacement parts
Ferrule, 1,58 mm, stainless steel, LC-20AD, LC-20AT, SIL-20AC, 228-16000-17, Old# 228-16000-10, 228-16000-84, SHIM
Catalog # CTSACTS-21015
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
- Description:Ferrule, 1,58 mm, stainless steel
- OEM manufacturer:SHIM
- OEM equivalent:228-16000-17, Old# 228-16000-10, 228-16000-84
- Model:LC-20AD, LC-20AT, SIL-20AC