Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 1 L
pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 1 L
Print
pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 1 L
Catalog # REGETB7001
Supplier:
REAGECON
pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 1 L
Catalog # REGETB7001
Supplier:
REAGECON
Expand image
Return to Top
pH Buffer Solution pH 7.00 ± 0.02 ,25°C (Coloured) 1 * 1 L
Add to cart
Price
52,50 €
Log in
for availability
Unit of Measure
1 * 1 L
-
+
Add to cart
Return Policy
Product Details & Documents