Order Entry
Export
Orders LinkContactUsLinkComponent
Human recombinant FGF-9

Human recombinant FGF-9

5 µG
Catalog # APOSBITP1051-5UG
Supplier:   Apollo Scientific
Human recombinant FGF-9
Catalog # APOSBITP1051-5UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant FGF-9

5 µG

CAS Number:  
Price
91,20 €
Log in for availability
1 * 5 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    5 µG
  • Cat. No.:
    APOSBITP1051-5UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FGF-9
  • Protein synonyms:
    Fibroblast Growth Factor-9
  • Species:
    Human

Specifications