0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Proteins and Peptides
Sheep Recombinant Prl-A
Sheep Recombinant Prl-A
10 µG
APOSBITP1255-10UG
:
Apollo Scientific
Sheep Recombinant Prl-A
APOSBITP1255-10UG
:
Apollo Scientific
:
Sheep Recombinant Prl-A
10 µG
:
91,20 €
1 * 10 µG
-
+
Pk:
10 µG
Cat. No.:
APOSBITP1255-10UG
Protein/peptide type:
Recombinant
Protein/peptide name:
Prl-A
Protein synonyms:
Prolactin Antagonist
Species:
Sheep