0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Proteins and Peptides
Human Recombinant Prl
Human Recombinant Prl
50 µG
APOSBITP1245-50UG
:
Apollo Scientific
Human Recombinant Prl
APOSBITP1245-50UG
:
Apollo Scientific
:
Human Recombinant Prl
50 µG
:
202,00 €
1 * 50 µG
-
+
Pk:
50 µG
Cat. No.:
APOSBITP1245-50UG
Protein/peptide type:
Recombinant
Protein/peptide name:
Prl
Protein synonyms:
Prolactin
Species:
Human