Order Entry
Export
Orders LinkContactUsLinkComponent
 

Human Recombinant IL-19

2 µG
Catalog # APOSBITP1181-2UG
Supplier:   Apollo Scientific
Human Recombinant IL-19
Catalog # APOSBITP1181-2UG
Supplier:  Apollo Scientific
CAS Number:  
Human Recombinant IL-19

2 µG

CAS Number:  
Price
91,20 €
Log in for availability
1 * 2 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    2 µG
  • Cat. No.:
    APOSBITP1181-2UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-19
  • Protein synonyms:
    Interleukin-19
  • Species:
    Human

Specifications