Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-2

Mouse Recombinant IL-2

20 µG
Catalog # APOSBITP1127-20UG
Supplier:   Apollo Scientific
Mouse Recombinant IL-2
Catalog # APOSBITP1127-20UG
Supplier:  Apollo Scientific
CAS Number:  
Mouse Recombinant IL-2

20 µG

CAS Number:  
Price
202,00 €
Log in for availability
1 * 20 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    20 µG
  • Cat. No.:
    APOSBITP1127-20UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-2
  • Protein synonyms:
    Interleukin-2
  • Species:
    Mouse

Specifications