Order Entry
Export
Orders LinkContactUsLinkComponent
 

Thiomorpholine-1,1-dioxide

Bottle, 1 g
Catalog # APOSOR13966-1G
Irritant
Supplier:   Apollo Scientific
Thiomorpholine-1,1-dioxide
Catalog # APOSOR13966-1G
Supplier:  Apollo Scientific
CAS Number:  39093-93-1
Thiomorpholine-1,1-dioxide

Bottle, 1 g

CAS Number:  39093-93-1
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Thiomorpholine 1,1-dioxide
  • CAS Number:39093-93-1
  • Formula:C₄H₉NO₂S
  • MDL Number:MFCD05861623
  • Molecular Weight:135.1848 g/mol
  • Boiling Pt:155 °C/1mmHg
  • Melting Pt:68…71 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications