Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,4-Diethynylbenzene 97%

Bottle, 1 g
Catalog # APOSOR13101-1G
Irritant
Supplier:   Apollo Scientific
1,4-Diethynylbenzene 97%
Catalog # APOSOR13101-1G
Supplier:  Apollo Scientific
CAS Number:  935-14-8
1,4-Diethynylbenzene 97%

Bottle, 1 g

CAS Number:  935-14-8
Price
40,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  p-Diethynylbenzene, 1,4-Diethylnylbenzene
  • CAS Number:935-14-8
  • Formula:C₁₀H₆
  • MDL Number:MFCD00078375
  • Molecular Weight:126.16 g/mol
  • Melting Pt:92 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications