Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Phenoxypropionic acid

Bottle, 25 g
Catalog # APOSOR13898-25G
Irritant
Supplier:   Apollo Scientific
3-Phenoxypropionic acid
Catalog # APOSOR13898-25G
Supplier:  Apollo Scientific
CAS Number:  7170-38-9
3-Phenoxypropionic acid

Bottle, 25 g

CAS Number:  7170-38-9
Price
232,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Penoxypropionic acid, 3-Phenoxypropanoic acid
  • CAS Number:7170-38-9
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002769
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:235…245 °C
  • Melting Pt:98…100 °C
  • Storage Temperature:Ambient
  • EINECS:230-518-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications