3-Phenoxypropionic acid
Bottle, 1 g
Catalog # APOSOR13898-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Penoxypropionic acid, 3-Phenoxypropanoic acid
- CAS Number:7170-38-9
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002769
- Molecular Weight:166.18 g/mol
- Boiling Pt:235…245 °C
- Melting Pt:98…100 °C
- Storage Temperature:Ambient
- EINECS:230-518-4
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



