2-Hydroxypyridine
Bottle, 25 g
APOSOR11197-25G
: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
- :142-08-5
- :C₅H₅NO
- :MFCD00006268
- :95.10 g/mol
- :280…281 °C
- :105…109 °C
- :Ambient
- :205-520-3
- :2811
- :6.1,III
- Pack type:Bottle
- Pk:25 g