0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,5-Dibromophenol
3,5-Dibromophenol 98%
3,5-Dibromophenol 98%
Bottle, 25 g
APOSOR11480-25G
:
Apollo Scientific
3,5-Dibromophenol 98%
APOSOR11480-25G
:
Apollo Scientific
:
626-41-5
3,5-Dibromophenol 98%
Bottle, 25 g
:
626-41-5
171,00 €
1 * 25 g
-
+
:
626-41-5
:
C₆H₄Br₂O
:
MFCD06411376
:
251.91 g/mol
:
79…83 °C
:
Ambient
:
2811
:
6.1,III
Pack type:
Bottle
Pk:
25 g