Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dibromophenol 98%

3,5-Dibromophenol 98%

Bottle, 100 g
Catalog # APOSOR11480-100G
Toxic
Supplier:   Apollo Scientific
3,5-Dibromophenol 98%
Catalog # APOSOR11480-100G
Supplier:  Apollo Scientific
CAS Number:  626-41-5
3,5-Dibromophenol 98%

Bottle, 100 g

CAS Number:  626-41-5
Price
545,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:626-41-5
  • Formula:C₆H₄Br₂O
  • MDL Number:MFCD06411376
  • Molecular Weight:251.91 g/mol
  • Melting Pt:79…83 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications