Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Phenylenediphosphonic acid

1,4-Phenylenediphosphonic acid

Bottle, 1 g
Catalog # APOSOR10672-1G
Corrosive
Irritant
Supplier:   Apollo Scientific
1,4-Phenylenediphosphonic acid
Catalog # APOSOR10672-1G
Supplier:  Apollo Scientific
CAS Number:  880-68-2
1,4-Phenylenediphosphonic acid

Bottle, 1 g

CAS Number:  880-68-2
Price
116,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1,4-Benzenediphosphonic acid
  • CAS Number:880-68-2
  • Formula:C₆H₈O₆P₂
  • MDL Number:MFCD00475582
  • Molecular Weight:238.07 g/mol
  • Melting Pt:>260 °C
  • Melting Pt:>260 °C
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications