Order Entry
Export
Orders LinkContactUsLinkComponent
4-Hydroxy-2-methoxybenzaldehyde 99.5+%

4-Hydroxy-2-methoxybenzaldehyde 99.5+%

Bottle, 25 g
Catalog # APOSOR0857-25G
Irritant
Supplier:   Apollo Scientific
4-Hydroxy-2-methoxybenzaldehyde 99.5+%
Catalog # APOSOR0857-25G
Supplier:  Apollo Scientific
CAS Number:  18278-34-7
4-Hydroxy-2-methoxybenzaldehyde 99.5+%

Bottle, 25 g

CAS Number:  18278-34-7
Price
142,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:18278-34-7
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00051964
  • Molecular Weight:152.15 g/mol
  • Melting Pt:156…161 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications