Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dichlorobenzhydrazide

3,5-Dichlorobenzhydrazide

Bottle, 5 g
Catalog # APOSOR0341-5G
Irritant
Supplier:   Apollo Scientific
3,5-Dichlorobenzhydrazide
Catalog # APOSOR0341-5G
Supplier:  Apollo Scientific
CAS Number:  62899-78-9
3,5-Dichlorobenzhydrazide

Bottle, 5 g

CAS Number:  62899-78-9
Price
145,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3,5-Dichlorobenzoic hydrazide
  • CAS Number:62899-78-9
  • Formula:C₇H₆Cl₂N₂O
  • MDL Number:MFCD00833411
  • Molecular Weight:205.04 g/mol
  • Melting Pt:205…207 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications