3,5-Dichlorobenzhydrazide
Bottle, 250 mg
Catalog # APOSOR0341-250MG
Supplier: Apollo Scientific
3,5-Dichlorobenzhydrazide
Catalog # APOSOR0341-250MG
Supplier: Apollo Scientific
CAS Number: 62899-78-9
Jump to:
Product Details & Documents
Synonyms: 3,5-Dichlorobenzoic hydrazide
- CAS Number:62899-78-9
- Formula:C₇H₆Cl₂N₂O
- MDL Number:MFCD00833411
- Molecular Weight:205.04 g/mol
- Melting Pt:205…207 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



