0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,5-Dichlorobenzhydrazide
3,5-Dichlorobenzhydrazide
3,5-Dichlorobenzhydrazide
3,5-Dichlorobenzhydrazide
Bottle, 1 g
APOSOR0341-1G
:
Apollo Scientific
3,5-Dichlorobenzhydrazide
APOSOR0341-1G
:
Apollo Scientific
:
62899-78-9
Expand image
3,5-Dichlorobenzhydrazide
Bottle, 1 g
Add to cart
:
62899-78-9
43,50 €
Unit of Measure
1 * 1 g
-
+
Add to cart
:
3,5-Dichlorobenzoic hydrazide
:
62899-78-9
:
C₇H₆Cl₂N₂O
:
MFCD00833411
:
205.04 g/mol
:
205…207 °C
:
Ambient
Pack type:
Bottle
Pk:
1 g
Cat. No.:
APOSOR0341-1G