2,3-Diphenylpiperazine
Bottle, 100 mg
Catalog # APOSOR01613-100MG
Supplier: Apollo Scientific
2,3-Diphenylpiperazine
Catalog # APOSOR01613-100MG
Supplier: Apollo Scientific
CAS Number: 143699-24-5
Jump to:
Product Details & Documents
- CAS Number:143699-24-5
- Formula:C₁₆H₁₈N₂
- MDL Number:MFCD06245414
- Molecular Weight:238.328 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:100 mg



