Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl propiolate

Methyl propiolate

Bottle, 5 g
Catalog # APOSOR0649-5G
Flammable
Irritant
Supplier:   Apollo Scientific
Methyl propiolate
Catalog # APOSOR0649-5G
Supplier:  Apollo Scientific
CAS Number:  922-67-8
Methyl propiolate

Bottle, 5 g

CAS Number:  922-67-8
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
  • CAS Number:922-67-8
  • Formula:C₄H₄O₂
  • MDL Number:MFCD00008572
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:103…105 °C
  • Density:0.945 g/cm³ (25 °C)
  • Flash Pt:10 °C
  • Storage Temperature:Ambient
  • EINECS:213-083-5
  • UN:3272
  • ADR:3,II

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications